Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroanisole 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
321-28-8
C7H7FO
126.13
MFCD00000316
JIXDOBAQOWOUPA-UHFFFAOYSA-N
2-fluoroanisole,o-fluoroanisole,benzene, 1-fluoro-2-methoxy,ortho-fluoroanisole,anisole, o-fluoro,1-fluoro-2-methoxy-benzene,paragos 390201,timtec-bb sbb040818,fr bo1 wln,fluoroanisole
67576
1-fluoro-2-methoxybenzene
COC1=CC=CC=C1F
321-28-8
C7H7FO
126.13
MFCD00000316
JIXDOBAQOWOUPA-UHFFFAOYSA-N
2-fluoroanisole,o-fluoroanisole,benzene, 1-fluoro-2-methoxy,ortho-fluoroanisole,anisole, o-fluoro,1-fluoro-2-methoxy-benzene,paragos 390201,timtec-bb sbb040818,fr bo1 wln,fluoroanisole
67576
1-fluoro-2-methoxybenzene
COC1=CC=CC=C1F
Infrared Spectrum Authentic
Refractive Index 1.4940 to 1.4970 (20°C, 589nm)
Color Orange
Quantity 100mL
Packaging Glass bottle
Density 1.1200g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4OCH3
Beilstein 06, II, 169
Formula Weight 126.13
Physical Form Crystalline Powder
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2-Fluoroanisole, 98%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-284-4

TSCA : TSCA