Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
1993-03-9
C6H6BFO2
139.92
MFCD00674013
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid,2-fluorobenzeneboronic acid,2-fluorophenyl boranediol,o-fluorophenylboronic acid,boronic acid, b-2-fluorophenyl,boronic acid, 2-fluorophenyl,2-fluoro-phenyl-boronic acid,o-fluoro-benzeneboronic acid,benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
B(C1=CC=CC=C1F)(O)O
1993-03-9
C6H6BFO2
139.92
MFCD00674013
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid,2-fluorobenzeneboronic acid,2-fluorophenyl boranediol,o-fluorophenylboronic acid,boronic acid, b-2-fluorophenyl,boronic acid, 2-fluorophenyl,2-fluoro-phenyl-boronic acid,o-fluoro-benzeneboronic acid,benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
B(C1=CC=CC=C1F)(O)O
Quantity 25g
Melting Point 100°C to 108°C
Beilstein 3030413
Formula Weight 139.92
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning