Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzenesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2905-21-7
C6H4ClFO2S
194.61
MFCD00042285
ZSZKAQCISWFDCQ-UHFFFAOYSA-N
2-fluorobenzene-1-sulfonyl chloride,2-fluorobenzenesulphonyl chloride,2-fluorobenzenesulfonylchloride,2-fluoro-benzenesulfonyl chloride,2-fluorophenylsulfonyl chloride,benzenesulfonyl chloride, 2-fluoro,chloro 2-fluorophenyl sulfone,pubchem5138,ksc205g6f
137761
2-fluorobenzenesulfonyl chloride
C1=CC=C(C(=C1)F)S(=O)(=O)Cl
2905-21-7
C6H4ClFO2S
194.61
MFCD00042285
ZSZKAQCISWFDCQ-UHFFFAOYSA-N
2-fluorobenzene-1-sulfonyl chloride,2-fluorobenzenesulphonyl chloride,2-fluorobenzenesulfonylchloride,2-fluoro-benzenesulfonyl chloride,2-fluorophenylsulfonyl chloride,benzenesulfonyl chloride, 2-fluoro,chloro 2-fluorophenyl sulfone,pubchem5138,ksc205g6f
137761
2-fluorobenzenesulfonyl chloride
C1=CC=C(C(=C1)F)S(=O)(=O)Cl
Boiling Point 140.0°C to 142.0°C (12.0mmHg)
Quantity 1g
Melting Point 26.0°C
Flash Point >110°C
Refractive Index 1.5350 to 1.5380
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Linear Formula FC6H4SO2Cl
Formula Weight 194.61
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger