Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluorobenzhydrazide, 98%

Click to view available options
Quantity:
10g
50g
250g
446-24-2
C7H7FN2O
154.144
MFCD00025112
YJCCKQQVXNNAAR-UHFFFAOYSA-N
2-fluorobenzhydrazide,2-fluorobenzoic hydrazide,o-fluorobenzhydrazide,2-fluorobenzoic acid hydrazide,2-fluorobenzoylhydrazine,2-fluorobenzenecarbohydrazide,pubchem3481,2-fluoro-benzohydrazide,acmc-209jyw,2-fluoranylbenzohydrazide
136288
2-fluorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)F
446-24-2
C7H7FN2O
154.144
MFCD00025112
YJCCKQQVXNNAAR-UHFFFAOYSA-N
2-fluorobenzhydrazide,2-fluorobenzoic hydrazide,o-fluorobenzhydrazide,2-fluorobenzoic acid hydrazide,2-fluorobenzoylhydrazine,2-fluorobenzenecarbohydrazide,pubchem3481,2-fluoro-benzohydrazide,acmc-209jyw,2-fluoranylbenzohydrazide
136288
2-fluorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)F
Quantity 10g
Melting Point 72°C to 74°C
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzhydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning