Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzoyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
393-52-2
C7H4ClFO
158.56
MFCD00000656
RAAGZOYMEQDCTD-UHFFFAOYSA-N
o-fluorobenzoyl chloride,benzoyl chloride, 2-fluoro,benzoyl chloride, o-fluoro,2-fluorobenzoylchloride,2-fluoro benzoyl chloride,unii-z4h017n76s,ccris 5914,fluorobenzoyl chloride,benzoyl chloride, fluoro,2-fluorobenzoic acid chloride
9808
2-fluorobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)F
393-52-2
C7H4ClFO
158.56
MFCD00000656
RAAGZOYMEQDCTD-UHFFFAOYSA-N
o-fluorobenzoyl chloride,benzoyl chloride, 2-fluoro,benzoyl chloride, o-fluoro,2-fluorobenzoylchloride,2-fluoro benzoyl chloride,unii-z4h017n76s,ccris 5914,fluorobenzoyl chloride,benzoyl chloride, fluoro,2-fluorobenzoic acid chloride
9808
2-fluorobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)F
Specific Gravity 1.32
Melting Point 4.0°C
Color Colorless
Density 1.3200g/mL
Flash Point 82°C
Quantity 25mL
Packaging Glass bottle
Refractive Index 1.5355 to 1.5375
Boiling Point 90.0°C to 92.0°C (15.0mmHg)
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4COCl
Beilstein 09, I, 136
Solubility Information Solubility in water: decomposes
Formula Weight 158.56
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzoyl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger