Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzylamine, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
89-99-6
C7H8FN
125.15
MFCD00008107
LRFWYBZWRQWZIM-UHFFFAOYSA-N
2-fluorobenzylamine,2-fluorophenyl methanamine,benzenemethanamine, 2-fluoro,o-fluorobenzylamine,benzylamine, o-fluoro,1-2-fluorophenyl methanamine,2-fluoro-benzylamine,2-fluorobenzyl amine,2-fluorophenyl methylamine,o-fluorobenzyl amine
66649
(2-fluorophenyl)methanamine
C1=CC=C(C(=C1)CN)F
89-99-6
C7H8FN
125.15
MFCD00008107
LRFWYBZWRQWZIM-UHFFFAOYSA-N
2-fluorobenzylamine,2-fluorophenyl methanamine,benzenemethanamine, 2-fluoro,o-fluorobenzylamine,benzylamine, o-fluoro,1-2-fluorophenyl methanamine,2-fluoro-benzylamine,2-fluorobenzyl amine,2-fluorophenyl methylamine,o-fluorobenzyl amine
66649
(2-fluorophenyl)methanamine
C1=CC=C(C(=C1)CN)F
Specific Gravity 1.09
Quantity 50g
Flash Point 67°C
Packaging Glass bottle
Refractive Index 1.5160 to 1.5190
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 73.0°C to 75.0°C (13.0mmHg)
Density 1.0900g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula FC6H4CH2NH2
Formula Weight 125.15
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2-Fluorobenzylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 201-957-9