Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobiphenyl 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
321-60-8
C12H9F
172.2
MFCD00000317
KLECYOQFQXJYBC-UHFFFAOYSA-N
2-fluorobiphenyl,2-fluoro-1,1'-biphenyl,o-fluorodiphenyl,1,1'-biphenyl, 2-fluoro,biphenyl, 2-fluoro,ortho-fluorodiphenyl,2-fluorodiphenyl,2-fluoro-biphenyl,ccris 1659,2-fluoro-1-phenylbenzene
67579
1-fluoro-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2F
321-60-8
C12H9F
172.2
MFCD00000317
KLECYOQFQXJYBC-UHFFFAOYSA-N
2-fluorobiphenyl,2-fluoro-1,1'-biphenyl,o-fluorodiphenyl,1,1'-biphenyl, 2-fluoro,biphenyl, 2-fluoro,ortho-fluorodiphenyl,2-fluorodiphenyl,2-fluoro-biphenyl,ccris 1659,2-fluoro-1-phenylbenzene
67579
1-fluoro-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2F
Boiling Point 248.0°C
Quantity 1g
Melting Point 74.0°C to 76.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5C6H4F
Beilstein 05,II,482
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material 2-Fluorobiphenyl, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementIF SWALLOWED: rinse mouth. Do NOT induce vomiting.IF ON SKIN: Wash with plenty of soap and water.If skin irritation occurs: Get medical advice/attention.IF IN EYES: Rinse cautiously with water for several minutes. Remove con

EINECSNumber : 206-290-7

RTECSNumber : DV5291000

TSCA : TSCA