Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Fluorophenethylamine, 97%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Specifications
Boiling Point | 64°C (0.6mmHg) |
Flash Point | 77°C |
Specific Gravity | 1.066 |
Refractive Index | 1.5100 to 1.512 |
Color | Orange to Yellow |
Quantity | 25g |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Density | 1.0660g/mL |
CAS Min % | 98.5 |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
TSCA : TSCA