Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenethyl bromide, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
91319-54-9
C8H8BrF
203.05
FQGDFWIQBCQXPS-UHFFFAOYSA-N
2-fluorophenethyl bromide,1-2-bromoethyl-2-fluorobenzene,1-2-bromo-ethyl-2-fluoro-benzene,benzene, 1-2-bromoethyl-2-fluoro,2-2-bromoethyl-1-fluorobenzene,1-2-bromoethyl-2-fluoro-benzene,2-fluorophenyl-ethylbromide,acmc-209wk4,2-fluorophenyl ethyl bromide,2-2-fluorophenyl-ethylbromide
2756964
1-(2-bromoethyl)-2-fluorobenzene
C1=CC=C(C(=C1)CCBr)F
91319-54-9
C8H8BrF
203.05
FQGDFWIQBCQXPS-UHFFFAOYSA-N
2-fluorophenethyl bromide,1-2-bromoethyl-2-fluorobenzene,1-2-bromo-ethyl-2-fluoro-benzene,benzene, 1-2-bromoethyl-2-fluoro,2-2-bromoethyl-1-fluorobenzene,1-2-bromoethyl-2-fluoro-benzene,2-fluorophenyl-ethylbromide,acmc-209wk4,2-fluorophenyl ethyl bromide,2-2-fluorophenyl-ethylbromide
2756964
1-(2-bromoethyl)-2-fluorobenzene
C1=CC=C(C(=C1)CCBr)F
Boiling Point 73°C to 75°C (20.0 Torr)
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 203.05
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Fluorophenethyl bromide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning