Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
Packaging:
Glass bottle
367-12-4
C6H5FO
112.1
MFCD00002155
HFHFGHLXUCOHLN-UHFFFAOYSA-N
o-fluorophenol,phenol, 2-fluoro,phenol, o-fluoro,1-fluoro-2-hydroxybenzene,2-fluorohydroxybenzene,fluorophenol,unii-i9ow1nly9r,2-fluoro-phenol,phenol, fluoro,ccris 1226
9707
2-fluorophenol
C1=CC=C(C(=C1)O)F
367-12-4
C6H5FO
112.1
MFCD00002155
HFHFGHLXUCOHLN-UHFFFAOYSA-N
o-fluorophenol,phenol, 2-fluoro,phenol, o-fluoro,1-fluoro-2-hydroxybenzene,2-fluorohydroxybenzene,fluorophenol,unii-i9ow1nly9r,2-fluoro-phenol,phenol, fluoro,ccris 1226
9707
2-fluorophenol
C1=CC=C(C(=C1)O)F
Refractive Index 1.5134 to 1.5154
Flash Point 46°C
Melting Point 16.1°C
Boiling Point 152°C
Specific Gravity 1.256
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.2560g/mL
Quantity 500g
Color Brown-Yellow to Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4OH
Beilstein 06, I, 97
Solubility Information Solubility in water: 37.7g/L (20°C)
Formula Weight 112.1
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Fluorophenol
Show More Show Less

GHS P Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Warning

Recommended Storage : this product darkens during storage