Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenylboronic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
1993-03-9
C6H6BFO2
139.92
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid,2-fluorobenzeneboronic acid,2-fluorophenyl boranediol,o-fluorophenylboronic acid,boronic acid, b-2-fluorophenyl,boronic acid, 2-fluorophenyl,2-fluoro-phenyl-boronic acid,o-fluoro-benzeneboronic acid,benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
B(C1=CC=CC=C1F)(O)O
1993-03-9
C6H6BFO2
139.92
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid,2-fluorobenzeneboronic acid,2-fluorophenyl boranediol,o-fluorophenylboronic acid,boronic acid, b-2-fluorophenyl,boronic acid, 2-fluorophenyl,2-fluoro-phenyl-boronic acid,o-fluoro-benzeneboronic acid,benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
B(C1=CC=CC=C1F)(O)O
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass Bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 139.92
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Fluorophenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning