Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoropropionic acid, 96%

Manufacturer:  Alfa Aesar™ H25841MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25841MD


  Check Availability
Add to cart

Specifications

Description

Applications
2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.

Solubility
Partly miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, strong bases.
Description & Specifications

Chemical Identifiers

6087-13-4
92.07
UN3265
C3H5FO2
MFCD06247714
2-Fluoropropionic acid

Specifications

82°C (180°F)
66° to 67°C (30mmHg)
6087-13-4
92.07
UN3265
96%
Acrid
250mg
C3H5FO2
MFCD06247714
2-Fluoropropionic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit