Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Fluoropyridine-4-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ 4475503

Quantity 1g

 View more versions of this product

Catalog No. AA4475503


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

402-65-3
141.1
2-Fluoroisonicotinic acid
C6H4FNO2
MFCD02181194
2-Fluoropyridine-4-carboxylic acid

Specifications

195° to 497°C
402-65-3
C6H4FNO2
MFCD02181194
2-Fluoropyridine-4-carboxylic acid
1g
Powder
141.1
2-Fluoroisonicotinic acid
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit