Alfa Aesar™ 2-Fluoropyridine-4-carboxylic acid, 98%

Manufacturer: Alfa Aesar™ 4475501

Quantity 0.25g

 View more versions of this product

Catalog No. AA4475501

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 402-65-3
    Molecular Formula C6H4FNO2
    Formula Weight 141.1
    MDL Number MFCD02181194
    Synonym 2-Fluoroisonicotinic acid
    Chemical Name or Material 2-Fluoropyridine-4-carboxylic acid

    Specifications

    Formula Weight 141.1
    Quantity 0.25g
    Physical Form Powder
    Molecular Formula C6H4FNO2
    MDL Number MFCD02181194
    Melting Point 195° to 497°C
    CAS 402-65-3
    Synonym 2-Fluoroisonicotinic acid
    Chemical Name or Material 2-Fluoropyridine-4-carboxylic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit