Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-4-methanol, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
131747-60-9
C6H6FNO
127.12
IAFXOPAZVVMMLE-UHFFFAOYSA-N
2-fluoropyridin-4-yl methanol,2-fluoro-4-pyridinemethanol,2-fluoro-pyridin-4-yl-methanol,4-pyridinemethanol, 2-fluoro,acmc-1c3lv,2-fluoropyridine-4-methanol,2-fluoro-4-hydroxymethylpyridine,4-pyridinemethanol, 2-fluoro-9ci
11332468
(2-fluoropyridin-4-yl)methanol
C1=CN=C(C=C1CO)F
131747-60-9
C6H6FNO
127.12
IAFXOPAZVVMMLE-UHFFFAOYSA-N
2-fluoropyridin-4-yl methanol,2-fluoro-4-pyridinemethanol,2-fluoro-pyridin-4-yl-methanol,4-pyridinemethanol, 2-fluoro,acmc-1c3lv,2-fluoropyridine-4-methanol,2-fluoro-4-hydroxymethylpyridine,4-pyridinemethanol, 2-fluoro-9ci
11332468
(2-fluoropyridin-4-yl)methanol
C1=CN=C(C=C1CO)F
Melting Point 61°C to 63°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 2-Fluoropyridine-4-methanol

GHS Signal Word: Warning