Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
50mL
250mL
Packaging:
Glass bottle
372-48-5
C5H4FN
97.09
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro,o-fluoropyridine,2-fluoro-pyridine,pyridine, fluoro,ccris 1725,2-fluoranylpyridine,fluoropyridine,2fluoropyridine,2-fluorpyridin,2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F
372-48-5
C5H4FN
97.09
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro,o-fluoropyridine,2-fluoro-pyridine,pyridine, fluoro,ccris 1725,2-fluoranylpyridine,fluoropyridine,2fluoropyridine,2-fluorpyridin,2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F
Specific Gravity 1.12
Infrared Spectrum Authentic
Quantity 5mL
Flash Point 24°C
Density 1.1200g/mL
Boiling Point 126.0°C (753.0 mmHg)
Color Colorless to Yellow
Refractive Index 1.4670 to 1.4690
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)F
Beilstein 20, I, 80
Formula Weight 97.09
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Fluoropyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning