Learn More
2-Fluoropyridine, 99%, Thermo Scientific Chemicals
Description
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.
Solubility
Soluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Density | 1.129 |
Boiling Point | 124°C to 126°C |
Flash Point | 24°C (75°F) |
Refractive Index | 1.467 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 1515 |
Solubility Information | Soluble in water. |
Formula Weight | 97.09 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 206-757-5 |
RTECSNumber | UT3900000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.