Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-fluorothiobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
75907-82-3
C7H6FNS
155.19
MFCD04973316
IEMYDFIJEIAQCO-UHFFFAOYSA-N
2-fluorothiobenzamide,2-fluorobenzothioamide,2-fluoro thiobenzamide,2-fluorobenzene-1-carbothioamide,amino 2-fluorophenyl methane-1-thione,2-fluoro-thiobenzamide,pubchem13565,benzenecarbothioamide,2-fluoro,benzenecarbothioamide, 2-fluoro,benzenecarbothioamide, 2-fluoro-9ci
2734821
2-fluorobenzenecarbothioamide
C1=CC=C(C(=C1)C(=S)N)F
75907-82-3
C7H6FNS
155.19
MFCD04973316
IEMYDFIJEIAQCO-UHFFFAOYSA-N
2-fluorothiobenzamide,2-fluorobenzothioamide,2-fluoro thiobenzamide,2-fluorobenzene-1-carbothioamide,amino 2-fluorophenyl methane-1-thione,2-fluoro-thiobenzamide,pubchem13565,benzenecarbothioamide,2-fluoro,benzenecarbothioamide, 2-fluoro,benzenecarbothioamide, 2-fluoro-9ci
2734821
2-fluorobenzenecarbothioamide
C1=CC=C(C(=C1)C(=S)N)F
Quantity 1g
Color Yellow
Melting Point 80°C to 84°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4CSNH2
Formula Weight 155.2
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Fluorothiobenzamide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning