Alfa Aesar™ 2-Fluorothiophenol, 97%

Manufacturer: Alfa Aesar™ B2499503

Quantity 1g

 View more versions of this product

Catalog No. AAB2499503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2557-78-0
    Molecular Formula C6H5FS
    Formula Weight 128.17
    MDL Number MFCD00041419
    UN Number UN3071
    Beilstein 2039768
    Synonym 2-Fluorobenzenethiol
    Chemical Name or Material 2-Fluorothiophenol

    Specifications

    Formula Weight 128.17
    MDL Number MFCD00041419
    UN Number UN3071
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H331-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Toxic if inhaled.
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Stench
    Boiling Point 164°C
    Molecular Formula C6H5FS
    CAS 2557-78-0
    Beilstein 2039768
    Synonym 2-Fluorobenzenethiol
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, TOXIC, FLAMMABLE, N.O.S.
    Flash Point 56°C (133°F)
    Quantity 1g
    Chemical Name or Material 2-Fluorothiophenol
    Assay 97%

    GHS H Statement
    H300-H310-H331-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Toxic if inhaled.
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit