Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorothiophenol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
2557-78-0
C6H5FS
128.164
MFCD00041419
WJTZZPVVTSDNJJ-UHFFFAOYSA-N
2-fluorothiophenol,o-fluorothiophenol,o-fluorobenzenethiol,2-fluorobenzene-1-thiol,fluorothiophenol,o-fluoro thiophenol,2-fluoro-thiophenol,2-fluoro-benzenethiol,2-fluorobenzene thiol,pubchem5428
520216
2-fluorobenzenethiol
C1=CC=C(C(=C1)F)S
2557-78-0
C6H5FS
128.164
MFCD00041419
WJTZZPVVTSDNJJ-UHFFFAOYSA-N
2-fluorothiophenol,o-fluorothiophenol,o-fluorobenzenethiol,2-fluorobenzene-1-thiol,fluorothiophenol,o-fluoro thiophenol,2-fluoro-thiophenol,2-fluoro-benzenethiol,2-fluorobenzene thiol,pubchem5428
520216
2-fluorobenzenethiol
C1=CC=C(C(=C1)F)S
Color Colorless to Yellow
Density 1.2030g/mL
Refractive Index 1.5530 to 1.5570 (20°C, 589nm)
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4SH
Formula Weight 128.17
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Fluorothiophenol, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger