Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furaldehyde, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Metal drum
98-01-1
C5H4O2
96.08
MFCD00003229
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furfural,2-furaldehyde,2-furancarboxaldehyde,furaldehyde,2-formylfuran,furfuraldehyde,fural,2-furanaldehyde,2-furancarbonal,2-furfural
7362
CHEBI:34768
furan-2-carbaldehyde
C1=COC(=C1)C=O
98-01-1
C5H4O2
96.08
MFCD00003229
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furfural,2-furaldehyde,2-furancarboxaldehyde,furaldehyde,2-formylfuran,furfuraldehyde,fural,2-furanaldehyde,2-furancarbonal,2-furfural
7362
CHEBI:34768
furan-2-carbaldehyde
C1=COC(=C1)C=O
Specific Gravity 1.16
Viscosity 1.49 mPa.s (25°C)
Flash Point 60°C
Boiling Point 159°C to 161°C
Melting Point -37°C
Refractive Index 1.5250 to 1.527
Water 0.2% max.
Residue after Evaporation 0.5% max.
Color Amber to Brown
Packaging Metal drum
Acidity 0.02 eq/L max.
Quantity 10L
Density 1.1600g/mL
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Merck Index 15,4333
Solubility Information Solubility in water: 83g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 96.08
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Furaldehyde
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sus

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f

GHS Signal Word: Danger

EINECSNumber : 202-627-7

Recommended Storage : This product darkens on contact with air