Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Furaldehyde, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acidity | 0.02 eq/L max. |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -37°C |
Color | Amber to Brown |
Density | 1.1600g/mL |
Boiling Point | 159°C to 161°C |
Flash Point | 60°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Flammable liquid and vapour. Toxic if swallowed. Harmful in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Sus |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to f |
EINECSNumber | 202-627-7 |
Recommended Storage | This product darkens on contact with air |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction