Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furaldehyde diethyl acetal, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
13529-27-6
C9H14O3
170.208
MFCD00003245
SEILDMUKBMYIEZ-UHFFFAOYSA-N
2-diethoxymethyl furan,2-furaldehyde diethyl acetal,furfural diethyl acetal,furan, 2-diethoxymethyl,unii-04b3gme3ij,2-furaldehydediethylacetal,2-diethoxymethyl-furan,furan, diethoxymethyl,2-furaldehyde diethylacetal,04b3gme3ij
83553
CHEBI:34769
2-(diethoxymethyl)furan
CCOC(C1=CC=CO1)OCC
13529-27-6
C9H14O3
170.208
MFCD00003245
SEILDMUKBMYIEZ-UHFFFAOYSA-N
2-diethoxymethyl furan,2-furaldehyde diethyl acetal,furfural diethyl acetal,furan, 2-diethoxymethyl,unii-04b3gme3ij,2-furaldehydediethylacetal,2-diethoxymethyl-furan,furan, diethoxymethyl,2-furaldehyde diethylacetal,04b3gme3ij
83553
CHEBI:34769
2-(diethoxymethyl)furan
CCOC(C1=CC=CO1)OCC
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Density 1g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 17, 278
Fieser 13,173; 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Formula Weight 170.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Furaldehyde diethyl acetal, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.