Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Furanylmethylene)malononitrile, 98+%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3237-22-7
C8H4N2O
114.13
CRUJHLHRJSNZIU-UHFFFAOYSA-N
furfurylidenemalononitrile,2-furan-2-ylmethylene malononitrile,furfural-malonitril,2-furanylmethylene malononitrile,malononitrile, furfurylidene,2-furan-2-ylmethylidene propanedinitrile,2-furfurylidenemalononitrile,furfural-malonitril german,1,1-dicyano-2-2-furyl ethylene,furan-2-ylmethylidene propanedinitrile
18606
2-(furan-2-ylmethylidene)propanedinitrile
C1=COC(=C1)C=C(C#N)C#N
3237-22-7
C8H4N2O
114.13
CRUJHLHRJSNZIU-UHFFFAOYSA-N
furfurylidenemalononitrile,2-furan-2-ylmethylene malononitrile,furfural-malonitril,2-furanylmethylene malononitrile,malononitrile, furfurylidene,2-furan-2-ylmethylidene propanedinitrile,2-furfurylidenemalononitrile,furfural-malonitril german,1,1-dicyano-2-2-furyl ethylene,furan-2-ylmethylidene propanedinitrile
18606
2-(furan-2-ylmethylidene)propanedinitrile
C1=COC(=C1)C=C(C#N)C#N
Melting Point 70°C to 74°C
Quantity 25g
Packaging Glass bottle
Assay Percent Range 98% min. (GC)
Formula Weight 114.13
Percent Purity 98+%
Chemical Name or Material (2-Furanylmethylene)malononitrile

GHS Signal Word: Warning