Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furoic acid hydrazide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3326-71-4
C5H6N2O2
126.115
MFCD00003235
SKTSVWWOAIAIKI-UHFFFAOYSA-N
2-furoic acid hydrazide,2-furoic hydrazide,2-furohydrazide,furoylhydrazide,2-furancarbohydrazide,2-furoylhydrazide,furoic acid, hydrazide,2-furoic acid, hydrazide,2-furoylhydrazine,2-furylcarbonylhydrazide
18731
furan-2-carbohydrazide
C1=COC(=C1)C(=O)NN
3326-71-4
C5H6N2O2
126.115
MFCD00003235
SKTSVWWOAIAIKI-UHFFFAOYSA-N
2-furoic acid hydrazide,2-furoic hydrazide,2-furohydrazide,furoylhydrazide,2-furancarbohydrazide,2-furoylhydrazide,furoic acid, hydrazide,2-furoic acid, hydrazide,2-furoylhydrazine,2-furylcarbonylhydrazide
18731
furan-2-carbohydrazide
C1=COC(=C1)C(=O)NN
Packaging Glass bottle
Color Beige
Melting Point 76°C to 80°C
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 18, 279
Formula Weight 126.11
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2-Furoic acid hydrazide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-046-2

RTECSNumber : LV1925000

TSCA : TSCA