Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furoylacetonitrile, 99%, Thermo Scientific™

Catalog Number AC355600010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
31909-58-7
C7H5NO2
135.12
MFCD00052210
RZNSHBXVTAHWPP-UHFFFAOYSA-N
2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
141671
3-(furan-2-yl)-3-oxopropanenitrile
O=C(CC#N)C1=CC=CO1
31909-58-7
C7H5NO2
135.12
MFCD00052210
RZNSHBXVTAHWPP-UHFFFAOYSA-N
2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
141671
3-(furan-2-yl)-3-oxopropanenitrile
O=C(CC#N)C1=CC=CO1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 76°C to 79°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 135.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Furoylacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit