Alfa Aesar™ 2-Furyldimethylsilanol sodium salt

Manufacturer: Alfa Aesar™ H30686MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH30686MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 879904-88-8
    Molecular Formula C6H9NaO2Si
    Formula Weight 164.21
    MDL Number MFCD08457663
    Chemical Name or Material 2-Furyldimethylsilanol sodium salt

    Specifications

    Formula Weight 164.21
    Molecular Formula C6H9NaO2Si
    MDL Number MFCD08457663
    CAS 879904-88-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈92°C
    Quantity 250mg
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Furyldimethylsilanol sodium salt

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit