Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Heptyne, 98+%

Click to view available options
Quantity:
2g
10g
50g
1119-65-9
C7H12
96.173
MFCD00039970
AMSFEMSYKQQCHL-UHFFFAOYSA-N
2-heptyne,1-methyl-2-butylacetylene,butylmethylacetylene,n-butylmethylacetylene,acmc-1brzr,n-c4h9c.$.cch3,2-heptyne,
14245
hept-2-yne
CCCCC#CC
1119-65-9
C7H12
96.173
MFCD00039970
AMSFEMSYKQQCHL-UHFFFAOYSA-N
2-heptyne,1-methyl-2-butylacetylene,butylmethylacetylene,n-butylmethylacetylene,acmc-1brzr,n-c4h9c.$.cch3,2-heptyne,
14245
hept-2-yne
CCCCC#CC
Odor Characteristic
Quantity 2g
Boiling Point 112°C to 113°C
Flash Point 11°C (52°F)
Linear Formula CH3(CH2)3C≡CCH3
UN Number UN3295
Beilstein 1733563
Formula Weight 96.17
Percent Purity ≥98%
Chemical Name or Material 2-Heptyne

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P261-P243-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.