Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexanone 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
591-78-6
C6H12O
100.16
MFCD00009482
QQZOPKMRPOGIEB-UHFFFAOYSA-N
2-hexanone,hexanone,2-oxohexane,propylacetone,n-butyl methyl ketone,butyl methyl ketone,methyl butyl ketone,methyl n-butyl ketone,hexanone-2,ketone, butyl methyl
11583
hexan-2-one
CCCCC(=O)C
591-78-6
C6H12O
100.16
MFCD00009482
QQZOPKMRPOGIEB-UHFFFAOYSA-N
2-hexanone,hexanone,2-oxohexane,propylacetone,n-butyl methyl ketone,butyl methyl ketone,methyl butyl ketone,methyl n-butyl ketone,hexanone-2,ketone, butyl methyl
11583
hexan-2-one
CCCCC(=O)C
Melting Point -57°C
Flash Point 23°C
Boiling Point 127°C
Specific Gravity 0.81
Density 0.8100g/mL
Refractive Index 1.3995 to 1.4015
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 2.5L
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)3COCH3
Beilstein 01, 689
Merck Index 15, 6107
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 100.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Hexanone, 98%
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-731-1

RTECSNumber : MP1400000

TSCA : TSCA