Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexanone, Reagent grade, 98%, Honeywell™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass Bottle
591-78-6
C6H12O
100.161
MFCD00009482
QQZOPKMRPOGIEB-UHFFFAOYSA-N
2-hexanone,hexanone,2-oxohexane,propylacetone,n-butyl methyl ketone,butyl methyl ketone,methyl butyl ketone,methyl n-butyl ketone,hexanone-2,ketone, butyl methyl
11583
hexan-2-one
CCCCC(=O)C
591-78-6
C6H12O
100.161
MFCD00009482
QQZOPKMRPOGIEB-UHFFFAOYSA-N
2-hexanone,hexanone,2-oxohexane,propylacetone,n-butyl methyl ketone,butyl methyl ketone,methyl butyl ketone,methyl n-butyl ketone,hexanone-2,ketone, butyl methyl
11583
hexan-2-one
CCCCC(=O)C

Reagent Grade, 98%

  • ≥95% for general laboratory use.
Packaging Glass Bottle
Quantity 50g
Boiling Point 127°C
Color Colorless
Density 0.812g/mL at 25°C
Melting Point -57°C
Refractive Index n20/D 1.401(lit.)
Flash Point 23°C (73.4°F)
Linear Formula CH3(CH2)3COCH3
UN Number UN1224
Beilstein 1737676
Formula Weight 100.16g/mol
Physical Form Liquid
Percent Purity 98%
Grade Reagent
Chemical Name or Material 2-Hexanone
Show More Show Less

P261-P281-P314

H226-H336-H361f-H372

DOTInformation : Class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all

EINECSNumber : 209-731-1

RTECSNumber : MP1400000