Alfa Aesar™ 2-Hexanoylfuran, 99%

Manufacturer: Alfa Aesar™ B2230614

Quantity 25g

 View more versions of this product

Catalog No. AAB2230614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14360-50-0
    Molecular Formula C10H14O2
    Formula Weight 166.22
    MDL Number MFCD00037354
    Beilstein 2956
    Synonym 2-Furyl n-pentyl ketone; n-Pentyl 2-furyl ketone
    Chemical Name or Material 2-Hexanoylfuran

    Specifications

    Formula Weight 166.22
    TSCA TSCA
    Molecular Formula C10H14O2
    MDL Number MFCD00037354
    Beilstein 2956
    Synonym 2-Furyl n-pentyl ketone; n-Pentyl 2-furyl ketone
    Flash Point 107°C (225°F)
    Odor Characteristic
    Boiling Point 125° to 127°C (15mmHg)
    Quantity 25g
    CAS 14360-50-0
    Chemical Name or Material 2-Hexanoylfuran
    Assay 99%

    Description

    Applications
    2-Hexanoylfuran is used as a food additives

    Solubility
    Difficult to mix.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit