Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hexanoylfuran, 99%

Click to view available options
Quantity:
5g
25g
14360-50-0
C10H14O2
166.22
MFCD00037354
YUAYWSBSIJVIBS-UHFFFAOYSA-N
2-hexanoylfuran,2-furyl pentyl ketone,1-hexanone, 1-2-furanyl,pentyl 2-furyl ketone,1-2-furyl-1-hexanone,1-furan-2-yl hexan-1-one,1-2-furyl hexanone,2-furyl n-pentyl ketone,1-2-furanyl-1-hexanone,furan, 2-hexanoyl
61738
1-(furan-2-yl)hexan-1-one
CCCCCC(=O)C1=CC=CO1
14360-50-0
C10H14O2
166.22
MFCD00037354
YUAYWSBSIJVIBS-UHFFFAOYSA-N
2-hexanoylfuran,2-furyl pentyl ketone,1-hexanone, 1-2-furanyl,pentyl 2-furyl ketone,1-2-furyl-1-hexanone,1-furan-2-yl hexan-1-one,1-2-furyl hexanone,2-furyl n-pentyl ketone,1-2-furanyl-1-hexanone,furan, 2-hexanoyl
61738
1-(furan-2-yl)hexan-1-one
CCCCCC(=O)C1=CC=CO1
Applications
2-Hexanoylfuran is used as a food additives

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Refractive Index 1.489
Flash Point 107°C (225°F)
Odor Characteristic
Density 0.994
Quantity 5g
Boiling Point 125°C to 127°C (15mmHg)
Beilstein 2956
Solubility Information Difficult to mix.
Formula Weight 166.22
Percent Purity 99%
Chemical Name or Material 2-Hexanoylfuran
Show More Show Less

EINECSNumber : 238-333-0

TSCA : Yes

Recommended Storage : Ambient temperatures