Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexyl-2,3,4,5,5-pentamethyl-3-imidazolinium-1-oxyl-methyl Sulfate, Free Radical, ACROS Organics™

Click to view available options
Quantity:
100mg
205526-63-2
C15H31N2O5S
351.482
MFCD00148923
GAXKZXPZERSGAW-UHFFFAOYSA-M
2-hexyl-3,2-hexyl-2,3,4,5,5-pentamethyl-3-imidazolinium-1-oxyl methyl sulfate, free radical
2723740
2-hexyl-3-$l^{1}-oxidanyl-1,2,4,4,5-pentamethylimidazol-1-ium;methyl sulfate
CCCCCCC1(N(C(C(=[N+]1C)C)(C)C)[O])C.COS(=O)(=O)[O-]
205526-63-2
C15H31N2O5S
351.482
MFCD00148923
GAXKZXPZERSGAW-UHFFFAOYSA-M
2-hexyl-3,2-hexyl-2,3,4,5,5-pentamethyl-3-imidazolinium-1-oxyl methyl sulfate, free radical
2723740
2-hexyl-3-$l^{1}-oxidanyl-1,2,4,4,5-pentamethylimidazol-1-ium;methyl sulfate
CCCCCCC1(N(C(C(=[N+]1C)C)(C)C)[O])C.COS(=O)(=O)[O-]
Polymerization reagent
Infrared Spectrum Authentic
Color Yellow
Melting Point 132°C to 136°C
Quantity 100mg
Formula Weight 351.49
Physical Form Fine Powder
Chemical Name or Material 2-Hexyl-2,3,4,5,5-pentamethyl-3-imidazolinium-1-oxyl methyl sulfate

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.