Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazino-2-imidazoline hydrobromide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
55959-84-7
C3H9BrN4
181.037
MFCD00012694
HIJRJVVSABWXKG-UHFFFAOYSA-N
2-hydrazino-2-imidazoline hydrobromide,2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide,2-hydrazino-delta-2-imidazoline hydrobromide,4,5-dihydroimidazol-2-ylhydrazine hydrobromide,4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide,2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide,4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide,4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide,4,5-dihydro-1h-imidazol-2-yl-hydrazine,acmc-1av1k
3085099
4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
C1CN=C(N1)NN.Br
55959-84-7
C3H9BrN4
181.037
MFCD00012694
HIJRJVVSABWXKG-UHFFFAOYSA-N
2-hydrazino-2-imidazoline hydrobromide,2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide,2-hydrazino-delta-2-imidazoline hydrobromide,4,5-dihydroimidazol-2-ylhydrazine hydrobromide,4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide,2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide,4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide,4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide,4,5-dihydro-1h-imidazol-2-yl-hydrazine,acmc-1av1k
3085099
4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
C1CN=C(N1)NN.Br
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 10g
Color White to Beige
Melting Point 181°C to 185°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 181.04
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Hydrazino-2-imidazoline hydrobromide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 259-921-3

TSCA : TSCA