Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazinopyridine 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4930-98-7
C5H7N3
109.13
MFCD00006249
NWELCUKYUCBVKK-UHFFFAOYSA-N
2-hydrazinylpyridine,2-hydrazinopyridine,2-pyridylhydrazine,2 1h-pyridinone, hydrazone,hydrazine, 2-pyridinyl,pyridine, 2-hydrazinyl,2-pyridylhydrazone,pyridin-2-yl-hydrazine,pyridine, 2-hydrazino,pyridin-2-yl hydrazine
78645
pyridin-2-ylhydrazine
C1=CC=NC(=C1)NN
4930-98-7
C5H7N3
109.13
MFCD00006249
NWELCUKYUCBVKK-UHFFFAOYSA-N
2-hydrazinylpyridine,2-hydrazinopyridine,2-pyridylhydrazine,2 1h-pyridinone, hydrazone,hydrazine, 2-pyridinyl,pyridine, 2-hydrazinyl,2-pyridylhydrazone,pyridin-2-yl-hydrazine,pyridine, 2-hydrazino,pyridin-2-yl hydrazine
78645
pyridin-2-ylhydrazine
C1=CC=NC(=C1)NN
Melting Point 44.5°C to 46.0°C
Boiling Point 185.0°C (140.0 mmHg)
Flash Point 110°C
Quantity 5g
Color Beige to White
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 22, I, 688
Formula Weight 109.13
Physical Form Low Melting Solid
Percent Purity 98%
Chemical Name or Material 2-Hydrazinopyridine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning