Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-1-naphthaldehyde, tech., Thermo Scientific™

Click to view available options
Quantity:
5g
100g
500g
708-06-5
C11H8O2
172.183
MFCD00004005
NTCCNERMXRIPTR-UHFFFAOYSA-N
2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
12819
2-hydroxynaphthalene-1-carbaldehyde
C1=CC=C2C(=C1)C=CC(=C2C=O)O
708-06-5
C11H8O2
172.183
MFCD00004005
NTCCNERMXRIPTR-UHFFFAOYSA-N
2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
12819
2-hydroxynaphthalene-1-carbaldehyde
C1=CC=C2C(=C1)C=CC(=C2C=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Color Yellow
Melting Point >77°C (on dry substance)
Infrared Spectrum Authentic
Linear Formula HOC10H6CHO
Quantity 5g
Formula Weight 172.18
Physical Form Crystalline Moist Powder
Percent Purity ≥93% (on dry substance)
Grade Technical
Water 25 to 60%
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde, tech.
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-902-0

RTECSNumber : QJ0300000

TSCA : TSCA

RUO – Research Use Only