Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-methylbutyric Acid, 95+%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
32 kg
Packaging:
Fiber Drum
Poly Bottle
3739-30-8
3739-30-8
Packaging Fiber Drum
Quantity 32 kg
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.