Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hydroxy-3-methoxybenzaldehyde, 99%

Click to view available options
Quantity:
25g
100g
500g
148-53-8
C8H8O3
152.149
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
148-53-8
C8H8O3
152.149
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde,o-vanillin,6-formylguaiacol,2-vanillin,o-vanilline,benzaldehyde, 2-hydroxy-3-methoxy,orthovanilline,2-hydroxy-m-anisaldehyde,6-formyl-2-methoxyphenol,3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(=C1O)C=O
Applications
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Sensitivity Air Sensitive
Melting Point 40°C to 42°C
Quantity 500g
Boiling Point 265°C to 266°C
Beilstein 471913
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-3-methoxybenzaldehyde

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 205-715-3

RTECSNumber : CU6530000

TSCA : Yes

Recommended Storage : Ambient temperatures