Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
33252-64-1
C6H3F3N2O3
208.096
MFCD00153193
JYXKHKBZLLIWEV-UHFFFAOYSA-N
2-hydroxy-3-nitro-5-trifluoromethyl pyridine,3-nitro-5-trifluoromethyl pyridin-2 1h-one,3-nitro-5-trifluoromethyl pyridin-2-ol,3-nitro-5-trifluoromethyl-2-pyridinone,2 1h-pyridinone,3-nitro-5-trifluoromethyl,5-trifluoromethyl-3-nitropyridin-2-ol,3-nitro-5-trifluoromethyl-1h-pyridin-2-one,2 1h-pyridinone, 3-nitro-5-trifluoromethyl,3-nitro-5-trifluoromethyl hydropyridin-2-one,pubchem9284
2775093
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
33252-64-1
C6H3F3N2O3
208.096
MFCD00153193
JYXKHKBZLLIWEV-UHFFFAOYSA-N
2-hydroxy-3-nitro-5-trifluoromethyl pyridine,3-nitro-5-trifluoromethyl pyridin-2 1h-one,3-nitro-5-trifluoromethyl pyridin-2-ol,3-nitro-5-trifluoromethyl-2-pyridinone,2 1h-pyridinone,3-nitro-5-trifluoromethyl,5-trifluoromethyl-3-nitropyridin-2-ol,3-nitro-5-trifluoromethyl-1h-pyridin-2-one,2 1h-pyridinone, 3-nitro-5-trifluoromethyl,3-nitro-5-trifluoromethyl hydropyridin-2-one,pubchem9284
2775093
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
Quantity 250mg
Melting Point 189°C to 190°C
Color White
UN Number UN2811
Formula Weight 208.09
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures