Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hydroxy-3-nitrobenzaldehyde, 98%

Click to view available options
Quantity:
1g
5g
5274-70-4
C7H5NO4
167.12
MFCD00041874
NUGOTBXFVWXVTE-UHFFFAOYSA-N
3-nitrosalicylaldehyde,salicylaldehyde, 3-nitro,benzaldehyde, 2-hydroxy-3-nitro,3-nitro-2-hydroxybenzaldehyde,3-nitrosalicaldehyde,pubchem7773,2-hydroxy-nitrobenzaldehyde,acmc-209l1g,3-nitrosalicylaidehyde,benzaldehyde, 2-hydroxynitro
78934
2-hydroxy-3-nitrobenzaldehyde
C1=CC(=C(C(=C1)[N+](=O)[O-])O)C=O
5274-70-4
C7H5NO4
167.12
MFCD00041874
NUGOTBXFVWXVTE-UHFFFAOYSA-N
3-nitrosalicylaldehyde,salicylaldehyde, 3-nitro,benzaldehyde, 2-hydroxy-3-nitro,3-nitro-2-hydroxybenzaldehyde,3-nitrosalicaldehyde,pubchem7773,2-hydroxy-nitrobenzaldehyde,acmc-209l1g,3-nitrosalicylaidehyde,benzaldehyde, 2-hydroxynitro
78934
2-hydroxy-3-nitrobenzaldehyde
C1=CC(=C(C(=C1)[N+](=O)[O-])O)C=O
Melting Point 109°C to 110°C
Quantity 5g
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-nitrobenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA