Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hydroxy-3-nitropyridine, 98%

Click to view available options
Quantity:
5g
25g
6332-56-5
C5H4N2O3
140.098
MFCD00006270
BOAFCICMVMFLIT-UHFFFAOYSA-N
2-hydroxy-3-nitropyridine,3-nitropyridin-2-ol,3-nitro-2-pyridinol,3-nitropyridin-2 1h-one,2 1h-pyridinone, 3-nitro,3-nitro-2-hydroxypyridine,pyridine, 2-hydroxy-3-nitro,3-nitro-1,2-dihydropyridin-2-one,3-nitro-2-oxopyridine,2-pyridinol, 3-nitro
22793
3-nitro-1H-pyridin-2-one
C1=CNC(=O)C(=C1)[N+](=O)[O-]
6332-56-5
C5H4N2O3
140.098
MFCD00006270
BOAFCICMVMFLIT-UHFFFAOYSA-N
2-hydroxy-3-nitropyridine,3-nitropyridin-2-ol,3-nitro-2-pyridinol,3-nitropyridin-2 1h-one,2 1h-pyridinone, 3-nitro,3-nitro-2-hydroxypyridine,pyridine, 2-hydroxy-3-nitro,3-nitro-1,2-dihydropyridin-2-one,3-nitro-2-oxopyridine,2-pyridinol, 3-nitro
22793
3-nitro-1H-pyridin-2-one
C1=CNC(=O)C(=C1)[N+](=O)[O-]
Quantity 25g
Melting Point ~220°C (decomposition)
Beilstein 124474
Formula Weight 140.1
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-nitropyridine

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H351

EINECSNumber : 228-709-2

RTECSNumber : UU7718000

TSCA : No

Recommended Storage : Ambient temperatures