Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-iodopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
13472-79-2
C5H4INO
220.99
ZDJUNNCVIDKJAN-UHFFFAOYSA-N
2-hydroxy-5-iodopyridine,5-iodopyridin-2-ol,5-iodopyridin-2 1h-one,5-iodo-pyridin-2-ol,5-iodo-2-pyridone,2 1h-pyridinone, 5-iodo,5-iodo-2-pyridinol,5-jod-2-pyridon,5-iodo-2-pyridol,5-iodopyridin-2-one
459500
5-iodo-1H-pyridin-2-one
C1=CC(=O)NC=C1I
13472-79-2
C5H4INO
220.99
ZDJUNNCVIDKJAN-UHFFFAOYSA-N
2-hydroxy-5-iodopyridine,5-iodopyridin-2-ol,5-iodopyridin-2 1h-one,5-iodo-pyridin-2-ol,5-iodo-2-pyridone,2 1h-pyridinone, 5-iodo,5-iodo-2-pyridinol,5-jod-2-pyridon,5-iodo-2-pyridol,5-iodopyridin-2-one
459500
5-iodo-1H-pyridin-2-one
C1=CC(=O)NC=C1I
Melting Point 186.7°C to 190.1°C
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 220.99
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-5-iodopyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning