Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-methoxyacetophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
705-15-7
C9H10O3
166.18
MFCD00008731
MLIBGOFSXXWRIY-UHFFFAOYSA-N
2'-hydroxy-5'-methoxyacetophenone,1-2-hydroxy-5-methoxyphenyl ethanone,2-hydroxy-5-methoxyacetophenone,1-2-hydroxy-5-methoxyphenyl ethan-1-one,ethanone, 1-2-hydroxy-5-methoxyphenyl,5-methoxy-2-hydroxyacetophenone,acetophenone, 2'-hydroxy-5'-methoxy,2'-hydroxy-5'-methoxyacetylphenone,1-2-hydroxy-5-methoxy-phenyl-ethanone,1-acetyl-2-hydroxy-5-methoxybenzene
69714
1-(2-hydroxy-5-methoxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)OC)O
705-15-7
C9H10O3
166.18
MFCD00008731
MLIBGOFSXXWRIY-UHFFFAOYSA-N
2'-hydroxy-5'-methoxyacetophenone,1-2-hydroxy-5-methoxyphenyl ethanone,2-hydroxy-5-methoxyacetophenone,1-2-hydroxy-5-methoxyphenyl ethan-1-one,ethanone, 1-2-hydroxy-5-methoxyphenyl,5-methoxy-2-hydroxyacetophenone,acetophenone, 2'-hydroxy-5'-methoxy,2'-hydroxy-5'-methoxyacetylphenone,1-2-hydroxy-5-methoxy-phenyl-ethanone,1-acetyl-2-hydroxy-5-methoxybenzene
69714
1-(2-hydroxy-5-methoxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)OC)O
Color Yellow
Melting Point 48°C to 51°C
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2'-Hydroxy-5'-methoxyacetophenone, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 211-882-3

TSCA : TSCA