Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
672-13-9
C8H8O3
152.15
MFCD00003332
FZHSPPYCNDYIKD-UHFFFAOYSA-N
5-methoxysalicylaldehyde,m-anisaldehyde, 6-hydroxy,benzaldehyde, 2-hydroxy-5-methoxy,salicylaldehyde, 5-methoxy,6-hydroxy-m-anisaldehyde,2-hydroxy-5-methoxy-benzaldehyde,5-methoxy-2-hydroxybenzaldehyde,2-hydroxy-5-methoxy benzaldehyde,2-formyl-4-methoxyphenol,pubchem2649
95695
2-hydroxy-5-methoxybenzaldehyde
COC1=CC(=C(C=C1)O)C=O
672-13-9
C8H8O3
152.15
MFCD00003332
FZHSPPYCNDYIKD-UHFFFAOYSA-N
5-methoxysalicylaldehyde,m-anisaldehyde, 6-hydroxy,benzaldehyde, 2-hydroxy-5-methoxy,salicylaldehyde, 5-methoxy,6-hydroxy-m-anisaldehyde,2-hydroxy-5-methoxy-benzaldehyde,5-methoxy-2-hydroxybenzaldehyde,2-hydroxy-5-methoxy benzaldehyde,2-formyl-4-methoxyphenol,pubchem2649
95695
2-hydroxy-5-methoxybenzaldehyde
COC1=CC(=C(C=C1)O)C=O
Specific Gravity 1.21
Flash Point >112°C
Melting Point 4°C
Boiling Point 103°C (2.5mmHg)
Density 1.2100g/mL
Refractive Index 1.5770 to 1.579
Quantity 5g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula HOC6H3(OCH3)CHO
Beilstein 08, 244
Formula Weight 152.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-589-