Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-methylbenzophenone, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1470-57-1
C14H12O2
212.248
MFCD00002379
OQERFUGURPLBQH-UHFFFAOYSA-N
2-hydroxy-5-methylbenzophenone,2-benzoyl-4-methylphenol,2-hydroxy-5-methylphenyl phenyl methanone,unii-85w615bmp7,methanone, 2-hydroxy-5-methylphenyl phenyl,2-hydroxy-5-methyl-benzophenone,2-hydroxy-5-methylphenyl-phenylmethanone,acmc-209cy0,dsstox_cid_27857,dsstox_rid_82610
73851
(2-hydroxy-5-methylphenyl)-phenylmethanone
CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2
1470-57-1
C14H12O2
212.248
MFCD00002379
OQERFUGURPLBQH-UHFFFAOYSA-N
2-hydroxy-5-methylbenzophenone,2-benzoyl-4-methylphenol,2-hydroxy-5-methylphenyl phenyl methanone,unii-85w615bmp7,methanone, 2-hydroxy-5-methylphenyl phenyl,2-hydroxy-5-methyl-benzophenone,2-hydroxy-5-methylphenyl-phenylmethanone,acmc-209cy0,dsstox_cid_27857,dsstox_rid_82610
73851
(2-hydroxy-5-methylphenyl)-phenylmethanone
CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2
Melting Point 83°C to 85°C
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H3(CH3)COC6H5
Beilstein 08, 177
Formula Weight 212.25
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Hydroxy-5-methylbenzophenone
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-002-1

TSCA : TSCA