Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-methylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
91914-06-6
C6H7NO
109.13
MFCD03092888
SOHMZGMHXUQHGE-UHFFFAOYSA-N
2-hydroxy-5-methylpyridine,5-methylpyridin-2-ol,2-hydroxy-5-picoline,5-methyl-2-pyridinol,5-methylpyridin-2 1h-one,5-methyl-2-pyridone,5-methyl-2 1h-pyridinone,2 1h-pyridinone, 5-methyl,2-pyridinol, 5-methyl,6-hydroxy-3-picoline
70482
5-methyl-1H-pyridin-2-one
CC1=CNC(=O)C=C1
91914-06-6
C6H7NO
109.13
MFCD03092888
SOHMZGMHXUQHGE-UHFFFAOYSA-N
2-hydroxy-5-methylpyridine,5-methylpyridin-2-ol,2-hydroxy-5-picoline,5-methyl-2-pyridinol,5-methylpyridin-2 1h-one,5-methyl-2-pyridone,5-methyl-2 1h-pyridinone,2 1h-pyridinone, 5-methyl,2-pyridinol, 5-methyl,6-hydroxy-3-picoline
70482
5-methyl-1H-pyridin-2-one
CC1=CNC(=O)C=C1
Melting Point 181°C to 183°C
Packaging Glass Bottle
Color Yellow
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 109.13
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Hydroxy-5-methylpyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.