Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, ACROS Organics™
Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
Specifications
Quantity | 250mg |
Melting Point | 31.0°C to 33.0°C |
Infrared Spectrum | Authentic |
Boiling Point | 82.0°C (60.0 mmHg) |
Packaging | Glass bottle |
Color | White to Yellow |
NMR | Consistent with structure |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98.5% min. (HPLC) |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning