Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-6'-methoxyacetophenone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
703-23-1
C9H10O3
166.176
MFCD00008732
UENLHUMCIOWYQN-UHFFFAOYSA-N
1-2-hydroxy-6-methoxyphenyl ethanone,2'-hydroxy-6'-methoxyacetophenone,2-hydroxy-6-methoxyacetophenone,1-2-hydroxy-6-methoxyphenyl ethan-1-one,ethanone, 1-2-hydroxy-6-methoxyphenyl,acetophenone, 2'-hydroxy-6'-methoxy,unii-m5vqg8968n,ethanone,1-2-hydroxy-6-methoxyphenyl,1-2-hydroxy-6-methoxy-phenyl-ethanone,1-acetyl-2-hydroxy-6-methoxybenzene
69709
1-(2-hydroxy-6-methoxyphenyl)ethanone
CC(=O)C1=C(C=CC=C1OC)O
703-23-1
C9H10O3
166.176
MFCD00008732
UENLHUMCIOWYQN-UHFFFAOYSA-N
1-2-hydroxy-6-methoxyphenyl ethanone,2'-hydroxy-6'-methoxyacetophenone,2-hydroxy-6-methoxyacetophenone,1-2-hydroxy-6-methoxyphenyl ethan-1-one,ethanone, 1-2-hydroxy-6-methoxyphenyl,acetophenone, 2'-hydroxy-6'-methoxy,unii-m5vqg8968n,ethanone,1-2-hydroxy-6-methoxyphenyl,1-2-hydroxy-6-methoxy-phenyl-ethanone,1-acetyl-2-hydroxy-6-methoxybenzene
69709
1-(2-hydroxy-6-methoxyphenyl)ethanone
CC(=O)C1=C(C=CC=C1OC)O
Quantity 5g
Assay Percent Range 97%
Formula Weight 166.18
Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone

EINECSNumber : 211-872-9