Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-p-naphthoquinone, 94%, pure, Acros Organics

Click to view available options
Quantity:
50g
83-72-7
C10H6O3
174.155
WVCHIGAIXREVNS-UHFFFAOYSA-N
lawsone,2-hydroxy-1,4-naphthoquinone,2-hydroxynaphthalene-1,4-dione,2-hydroxynaphthoquinone,henna,mehendi,hana,lawson,mendi,flower of paradise
6755
4-hydroxynaphthalene-1,2-dione
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O
83-72-7
C10H6O3
174.155
WVCHIGAIXREVNS-UHFFFAOYSA-N
lawsone,2-hydroxy-1,4-naphthoquinone,2-hydroxynaphthalene-1,4-dione,2-hydroxynaphthoquinone,henna,mehendi,hana,lawson,mendi,flower of paradise
6755
4-hydroxynaphthalene-1,2-dione
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O
Quantity 50g
Infrared Spectrum Authentic
Color Yellow to Orange
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Beilstein 08, 300
Fieser 01,484
Merck Index 15, 5448
Formula Weight 174.16
Physical Form Crystalline Powder
Percent Purity ≥92%
Grade Pure
Chemical Name or Material 2-Hydroxy-p-naphthoquinone, tech.
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.