Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxyacetophenone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
118-93-4
C8H8O2
136.15
MFCD00002219
JECYUBVRTQDVAT-UHFFFAOYSA-N
2'-hydroxyacetophenone,2-acetylphenol,1-2-hydroxyphenyl ethanone,o-hydroxyacetophenone,o-acetylphenol,o-hydroxyphenyl methyl ketone,ethanone, 1-2-hydroxyphenyl,1-2-hydroxyphenyl ethan-1-one,acetophenone, 2'-hydroxy,2-hydroxyphenyl methyl ketone
8375
1-(2-hydroxyphenyl)ethanone
CC(=O)C1=CC=CC=C1O
118-93-4
C8H8O2
136.15
MFCD00002219
JECYUBVRTQDVAT-UHFFFAOYSA-N
2'-hydroxyacetophenone,2-acetylphenol,1-2-hydroxyphenyl ethanone,o-hydroxyacetophenone,o-acetylphenol,o-hydroxyphenyl methyl ketone,ethanone, 1-2-hydroxyphenyl,1-2-hydroxyphenyl ethan-1-one,acetophenone, 2'-hydroxy,2-hydroxyphenyl methyl ketone
8375
1-(2-hydroxyphenyl)ethanone
CC(=O)C1=CC=CC=C1O
Packaging Glass bottle
Color Brown to Yellow
Infrared Spectrum Authentic
Quantity 5g
Density 1.1310g/mL
Refractive Index 1.5580 to 1.5600 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4COCH3
Beilstein 08, 85
Formula Weight 136.15
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2'-Hydroxyacetophenone, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-288-0

RTECSNumber : AM8575000

TSCA : TSCA